Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272639.2 | NP_001259568.1 | 1 - 2 | 329 | 72 | |
NM_078607.3 | NP_511162.1 | 1 - 2 | 329 | 72 | ||
GenBank mRNA | BT016120.1 | - | 1 - 2 | 323 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |