Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pkcdelta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144719.2 | NP_001138191.1 | 10 - 11 | 1735 | 65 | |
NM_078616.4 | NP_511171.3 | 10 - 11 | 1911 | 65 | ||
NM_132569.3 | NP_572797.3 | 3 - 4 | 4980 | 65 | ||
GenBank mRNA | BT021275.1 | - | 9 - 10 | 1667 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |