Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258777.1 | NP_001245706.1 | 4 - 5 | 4033 | 76 | |
NM_001258778.2 | NP_001245707.1 | 7 - 8 | 1848 | 76 | ||
NM_001298390.1 | NP_001285319.1 | 5 - 6 | 3981 | 76 | ||
NM_078640.4 | NP_523364.2 | 4 - 5 | 1601 | 76 | ||
NM_167489.2 | NP_727941.2 | 5 - 6 | 1737 | 76 | ||
NM_167490.3 | NP_727942.1 | 4 - 5 | 1669 | 76 | ||
NM_167491.2 | NP_727943.1 | 4 - 5 | 1801 | 76 | ||
NM_176741.3 | NP_788914.2 | 6 - 7 | 1807 | 76 | ||
GenBank mRNA | AY118437.1 | - | 4 - 5 | 1605 | 76 | |
L35604.1 | - | 4 - 5 | 1631 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |