Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Dm01813738_m1
See other PARA GE Assays ›
Gene Symbol:
para
Gene Name:
paralytic
Gene Aliases:
CG9907, DmNa[[V]], DmNa[[v]], DmNa[[v]]1, DmNav, DmNav1, Dmel\CG9907, Dmel_CG9907, Nav1, Ocd, Para, Shu, Shudderer, VSSC, bas, bss, l(1)14Da, l(1)ESHS48, lincRNA.S9469, olfD, par, sbl, sbl-1
Chromosome Location:
Chr.X: 16455027 - 16533096 on Build Release_6_plus_ISO1_MT
Species:
Drosophila melanogaster
Species Specific ID (Flybase ID):
FBgn0285944
Assay ID
Dm01813738_m1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: para
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001042815.2
NP_001036280.1
29 - 30
5346
184
NM_001042816.2
NP_001036281.1
29 - 30
5346
184
NM_001144733.2
NP_001138205.1
28 - 29
5295
184
NM_001201671.1
NP_001188600.1
29 - 30
5277
184
NM_001201672.1
NP_001188601.1
29 - 30
5277
184
NM_001201673.1
NP_001188602.1
28 - 29
5214
184
NM_001201674.1
NP_001188603.1
28 - 29
5244
184
NM_001201675.1
NP_001188604.1
28 - 29
5190
184
NM_001201676.1
NP_001188605.1
29 - 30
5253
184
NM_001201677.1
NP_001188606.1
29 - 30
5181
184
NM_001201678.1
NP_001188607.1
28 - 29
5214
184
NM_001201679.1
NP_001188608.1
29 - 30
5253
184
NM_001201680.1
NP_001188609.1
28 - 29
5253
184
NM_001201681.1
NP_001188610.1
28 - 29
5229
184
NM_001201682.1
NP_001188611.1
29 - 30
5316
184
NM_001201683.1
NP_001188612.1
29 - 30
5181
184
NM_001201684.1
NP_001188613.1
28 - 29
5190
184
NM_001201685.1
NP_001188614.1
28 - 29
5253
184
NM_001201686.1
NP_001188615.1
29 - 30
5283
184
NM_001201687.1
NP_001188616.1
29 - 30
5307
184
NM_001201688.1
NP_001188617.1
28 - 29
5244
184
NM_001201689.1
NP_001188618.1
28 - 29
5259
184
NM_001201690.1
NP_001188619.1
29 - 30
5328
184
NM_001201691.1
NP_001188620.1
29 - 30
5367
184
NM_001201692.1
NP_001188621.1
29 - 30
5220
184
NM_001201693.1
NP_001188622.1
29 - 30
5205
184
NM_001201694.1
NP_001188623.1
29 - 30
5211
184
NM_001201695.2
NP_001188624.1
29 - 30
5205
184
NM_001201696.1
NP_001188625.1
27 - 28
5211
184
NM_001201697.1
NP_001188626.1
29 - 30
5277
184
NM_001201698.1
NP_001188627.1
28 - 29
5283
184
NM_001201699.1
NP_001188628.1
27 - 28
5220
184
NM_001201700.1
NP_001188629.1
28 - 29
5274
184
NM_001201701.1
NP_001188630.1
27 - 28
5211
184
NM_001201702.1
NP_001188631.1
28 - 29
5349
184
NM_001201703.3
NP_001188632.1
29 - 30
5767
184
NM_001201704.1
NP_001188633.1
27 - 28
5286
184
NM_001201705.1
NP_001188634.1
28 - 29
5202
184
NM_001201706.1
NP_001188635.1
29 - 30
5382
184
NM_001201707.1
NP_001188636.1
29 - 30
5292
184
NM_001201708.1
NP_001188637.1
28 - 29
5244
184
NM_001201709.1
NP_001188638.1
28 - 29
5220
184
NM_001201710.1
NP_001188639.1
25 - 26
5104
184
NM_001201711.1
NP_001188640.1
27 - 28
5154
184
NM_001201712.1
NP_001188641.1
26 - 27
5121
184
NM_001201713.1
NP_001188642.1
24 - 25
4462
184
NM_001201714.1
NP_001188643.1
24 - 25
4438
184
NM_001201715.1
NP_001188644.1
27 - 28
5139
184
NM_001201716.1
NP_001188645.1
28 - 29
5250
184
NM_001201717.1
NP_001188646.1
28 - 29
5349
184
NM_001201718.1
NP_001188647.1
27 - 28
5286
184
NM_001201719.1
NP_001188648.1
28 - 29
5349
184
NM_001201720.1
NP_001188649.1
27 - 28
5151
184
NM_001201721.1
NP_001188650.1
23 - 24
4209
184
NM_001201722.1
NP_001188651.1
27 - 28
5121
184
NM_001201723.1
NP_001188652.1
27 - 28
5162
184
NM_001272690.1
NP_001259619.1
29 - 30
5346
184
NM_001298403.1
NP_001285332.1
29 - 30
5499
184
NM_001298404.1
NP_001285333.1
27 - 28
5118
184
NM_078647.4
NP_523371.2
29 - 30
5346
184
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Drosophila melanogaster
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
mechanosensory behavior
response to mechanical stimulus
neuronal action potential
sodium ion transmembrane transport
male courtship behavior, veined wing generated song production
regulation of postsynaptic membrane potential
membrane depolarization during action potential
voltage-gated sodium channel activity
cation channel activity
sodium channel activity
calcium ion binding
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0