Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103531.2 | NP_001097001.1 | 3 - 4 | 503 | 64 | |
NM_078648.4 | NP_523372.2 | 2 - 3 | 742 | 64 | ||
GenBank mRNA | AY061256.1 | - | 2 - 3 | 742 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |