Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: if | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169306.2 | NP_001162777.1 | 5 - 6 | 779 | 81 | |
NM_001169307.1 | NP_001162778.1 | 5 - 6 | 914 | 81 | ||
NM_078654.2 | NP_523378.2 | 5 - 6 | 820 | 81 | ||
NM_167544.2 | NP_728021.2 | 5 - 6 | 820 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |