Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B-H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078662.3 | NP_523386.1 | 1 - 2 | 1331 | 94 | |
GenBank mRNA | BT022144.1 | - | 1 - 2 | 1238 | 94 | |
M82884.1 | - | 1 - 2 | 1332 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |