Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: beta-Spec | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272731.1 | NP_001259660.1 | 7 - 8 | 7102 | 70 | |
NM_001272732.1 | NP_001259661.1 | 7 - 8 | 7280 | 70 | ||
NM_078664.3 | NP_523388.1 | 6 - 7 | 7055 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |