Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhc16F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078670.3 | NP_523394.1 | 4 - 5 | 1654 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |