Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: run | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298572.1 | NP_001285501.1 | 2 - 3 | 1634 | 81 | |
NM_078700.4 | NP_523424.2 | 1 - 2 | 963 | 81 | ||
GenBank mRNA | BT029257.1 | - | 1 - 2 | 947 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |