Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ets21C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103577.3 | NP_001097047.1 | 2 - 3 | 712 | 72 | |
NM_001298618.1 | NP_001285547.1 | 2 - 3 | 1532 | 72 | ||
NM_001298619.1 | NP_001285548.1 | 2 - 3 | 2001 | 72 | ||
NM_078721.2 | NP_523445.1 | 2 - 3 | 1532 | 72 | ||
GenBank mRNA | BT031000.1 | - | 2 - 3 | 1532 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Drosophila melanogaster |