Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ser12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078732.2 | NP_523456.1 | 1 | 630 | 90 | |
GenBank mRNA | BT023348.1 | - | 1 | 630 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Drosophila melanogaster |