Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rab5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001272996.1 | NP_001259925.1 | 5 - 6 | 956 | 138 | |
NM_001272998.1 | NP_001259927.1 | 5 - 6 | 946 | 138 | ||
NM_078733.4 | NP_523457.1 | 5 - 6 | 929 | 138 | ||
NM_164475.2 | NP_722795.1 | 5 - 6 | 936 | 138 | ||
NM_164476.2 | NP_722796.1 | 5 - 6 | 1042 | 138 | ||
NM_164477.3 | NP_722797.1 | 5 - 6 | 1238 | 138 | ||
NM_164478.2 | NP_722798.1 | 5 - 6 | 910 | 138 | ||
GenBank mRNA | AB035353.1 | - | 5 - 6 | 944 | 137 | |
AB035671.1 | - | 5 - 6 | 915 | 138 | ||
AY060343.1 | - | 5 - 6 | 1214 | 138 | ||
AY081181.1 | - | 5 - 6 | 898 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |