Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078735.6 | NP_523459.3 | 1 - 2 | 869 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Drosophila melanogaster |