Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gr23a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_078735.6 | NP_523459.3 | 2 - 3 | 1035 | 98 | |
NM_175951.2 | NP_787965.1 | 2 - 3 | 1047 | 98 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Recommended Assay Drosophila melanogaster |