Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lilli | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144300.2 | NP_001137772.1 | 6 - 7 | 4396 | 72 | |
NM_001144301.1 | NP_001137773.1 | 7 - 8 | 4736 | 72 | ||
NM_001144302.2 | NP_001137774.1 | 6 - 7 | 4505 | 72 | ||
NM_001144303.1 | NP_001137775.1 | 8 - 9 | 6040 | 72 | ||
NM_001273044.1 | NP_001259973.1 | 7 - 8 | 6193 | 72 | ||
NM_001347818.1 | NP_001334729.1 | 7 - 8 | 6193 | 72 | ||
NM_078740.3 | NP_523464.1 | 8 - 9 | 6956 | 72 | ||
NM_164516.2 | NP_722863.1 | 9 - 10 | 7124 | 72 | ||
NM_164517.1 | NP_722864.1 | 8 - 9 | 5845 | 72 | ||
GenBank mRNA | AF289034.1 | - | 9 - 10 | 7101 | 72 | |
AF293971.1 | - | 8 - 9 | 6930 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |