Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014463.4 | NP_001014463.2 | 15 - 16 | 4578 | 61 | |
NM_001038783.3 | NP_001033872.2 | 14 - 15 | 5005 | 61 | ||
NM_001144308.3 | NP_001137780.2 | 16 - 17 | 4384 | 61 | ||
NM_001347756.1 | NP_001334730.1 | 15 - 16 | 4437 | 61 | ||
NM_164540.4 | NP_722912.3 | 15 - 16 | 4768 | 61 | ||
NM_164541.6 | NP_722913.3 | 15 - 16 | 4441 | 61 | ||
NM_164542.5 | NP_722914.3 | 15 - 16 | 4437 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Drosophila melanogaster |