Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atet | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103608.2 | NP_001097078.1 | 6 - 7 | 2708 | 71 | |
NM_001103609.3 | NP_001097079.1 | 6 - 7 | 3045 | 71 | ||
NM_078747.3 | NP_523471.2 | 6 - 7 | 3045 | 71 | ||
NM_164580.2 | NP_722971.1 | 6 - 7 | 2710 | 71 | ||
GenBank mRNA | BT029941.1 | - | 6 - 7 | 2708 | 71 | |
BT029944.1 | - | 6 - 7 | 2708 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |