Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gart | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001298769.1 | NP_001285698.1 | 4 - 5 | 1494 | 74 | |
NM_078773.3 | NP_523497.2 | 4 - 5 | 1494 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |