Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sick | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103713.4 | NP_001097183.1 | 13 - 14 | 6239 | 52 | |
NM_001273674.1 | NP_001260603.1 | 15 - 16 | 7102 | 52 | ||
NM_001273675.1 | NP_001260604.1 | 5 - 6 | 4395 | 52 | ||
NM_001273676.1 | NP_001260605.1 | 13 - 14 | 6355 | 52 | ||
NM_001273677.1 | NP_001260606.1 | 14 - 15 | 8121 | 52 | ||
NM_001273678.1 | NP_001260607.1 | 14 - 15 | 8807 | 52 | ||
NM_001273679.1 | NP_001260608.1 | 10 - 11 | 4572 | 52 | ||
NM_001273680.2 | NP_001260609.1 | 16 - 17 | 7034 | 52 | ||
NM_001273681.1 | NP_001260610.1 | 13 - 14 | 6560 | 52 | ||
NM_136167.3 | NP_610011.2 | 5 - 6 | 4294 | 52 | ||
NM_206007.5 | NP_995729.2 | 15 - 16 | 7007 | 52 | ||
NM_206008.4 | NP_995730.2 | 15 - 16 | 8974 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |