Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bwa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273687.2 | NP_001260616.1 | 1 - 2 | 509 | 58 | |
NM_136176.4 | NP_610020.1 | 1 - 2 | 509 | 58 | ||
GenBank mRNA | AY071232.1 | - | 1 - 2 | 542 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |