Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299173.1 | NP_001286102.1 | 6 - 7 | 1308 | 63 | |
NM_136189.4 | NP_610033.1 | 6 - 7 | 1275 | 63 | ||
GenBank mRNA | AY060391.1 | - | 6 - 7 | 1251 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |