Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: del | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273736.1 | NP_001260665.1 | 1 - 2 | 218 | 69 | |
NM_001273737.1 | NP_001260666.1 | 1 - 2 | 347 | 69 | ||
NM_136247.3 | NP_610091.2 | 1 - 2 | 218 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |