Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nipped-A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103722.3 | NP_001097192.2 | 11 - 12 | 2492 | 65 | |
NM_001316406.1 | NP_001303335.1 | 11 - 12 | 2492 | 65 | ||
GenBank mRNA | DQ352451.1 | - | 11 - 12 | 2518 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |