Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sced | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136375.2 | NP_610219.2 | 1 | 3137 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |