Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_136395.4 | NP_610239.1 | 2 - 3 | 268 | 84 | |
GenBank mRNA | BT023690.1 | - | 2 - 3 | 252 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |