Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273852.1 | NP_001260781.1 | 15 - 16 | 4951 | 90 | |
NM_136455.3 | NP_610299.2 | 15 - 16 | 4835 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |