Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273852.1 | NP_001260781.1 | 17 - 18 | 5273 | 146 | |
NM_136455.3 | NP_610299.2 | 17 - 18 | 5157 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |