Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001273852.1 | NP_001260781.1 | 29 - 30 | 10028 | 71 | |
NM_136455.3 | NP_610299.2 | 29 - 30 | 9912 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |