Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vha44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144207.2 | NP_001137679.1 | 9 - 10 | 2428 | 66 | |
NM_057918.5 | NP_477266.1 | 7 - 8 | 1084 | 66 | ||
NM_134313.4 | NP_599140.1 | 7 - 8 | 1139 | 66 | ||
NM_166163.3 | NP_725564.1 | 8 - 9 | 1301 | 66 | ||
NM_166164.5 | NP_725565.3 | 9 - 10 | 2483 | 66 | ||
GenBank mRNA | AY061038.1 | - | 7 - 8 | 1118 | 66 | |
AY102676.1 | - | 7 - 8 | 1084 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |