Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Syn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079038.5 | NP_523762.2 | 7 - 8 | 1312 | 74 | |
NM_166179.4 | NP_725587.1 | 9 - 10 | 1800 | 74 | ||
NM_166180.4 | NP_725588.1 | 9 - 10 | 1604 | 74 | ||
NM_176201.3 | NP_788381.1 | 8 - 9 | 1477 | 74 | ||
GenBank mRNA | AY094895.1 | - | 9 - 10 | 1602 | 74 | |
BT132953.1 | - | 8 - 9 | 1477 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Drosophila melanogaster |