Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299564.1 | NP_001286493.1 | 3 - 4 | 487 | 63 | |
NM_001299565.1 | NP_001286494.1 | 4 - 5 | 640 | 63 | ||
NM_057848.3 | NP_477196.1 | 4 - 5 | 1066 | 63 | ||
NM_166184.2 | NP_725594.1 | 4 - 5 | 841 | 63 | ||
NM_166185.2 | NP_725595.1 | 4 - 5 | 542 | 63 | ||
GenBank mRNA | AY060397.1 | - | 4 - 5 | 1048 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |