Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299567.1 | NP_001286496.1 | 2 - 3 | 629 | 64 | |
NM_001299568.1 | NP_001286497.1 | 4 - 5 | 1132 | 64 | ||
NM_137302.3 | NP_611146.1 | 3 - 4 | 997 | 64 | ||
NM_166187.2 | NP_725600.1 | 2 - 3 | 595 | 64 | ||
GenBank mRNA | AY069501.1 | - | 3 - 4 | 987 | 64 | |
BT125004.1 | - | 2 - 3 | 425 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |