Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Picot | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299567.1 | NP_001286496.1 | 3 - 4 | 1192 | 90 | |
NM_001299568.1 | NP_001286497.1 | 5 - 6 | 1695 | 90 | ||
NM_137302.3 | NP_611146.1 | 4 - 5 | 1560 | 90 | ||
NM_166187.2 | NP_725600.1 | 3 - 4 | 1158 | 90 | ||
GenBank mRNA | AY069501.1 | - | 4 - 5 | 1550 | 90 | |
BT125004.1 | - | 3 - 4 | 988 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |