Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qkr54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274121.1 | NP_001261050.1 | 4 - 5 | 988 | 73 | |
NM_079048.4 | NP_523772.1 | 4 - 5 | 1353 | 73 | ||
NM_166230.3 | NP_725679.1 | 4 - 5 | 998 | 73 | ||
NM_166231.3 | NP_725680.2 | 4 - 5 | 988 | 73 | ||
GenBank mRNA | AF096272.1 | - | 4 - 5 | 952 | 73 | |
AY095201.1 | - | 4 - 5 | 980 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |