Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: qkr54B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274121.1 | NP_001261050.1 | 7 - 8 | 1600 | 102 | |
NM_079048.4 | NP_523772.1 | 7 - 8 | 1965 | 102 | ||
NM_166230.3 | NP_725679.1 | 7 - 8 | 1610 | 102 | ||
NM_166231.3 | NP_725680.2 | 7 - 8 | 1600 | 102 | ||
GenBank mRNA | AF096272.1 | - | 7 - 8 | 1564 | 102 | |
AY095201.1 | - | 7 - 8 | 1592 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Drosophila melanogaster Inventoried |