Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: grh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259472.1 | NP_001246401.1 | 12 - 13 | 2251 | 88 | |
NM_001299622.1 | NP_001286551.1 | 12 - 13 | 5302 | 88 | ||
NM_057494.4 | NP_476842.2 | 11 - 12 | 3840 | 88 | ||
NM_057495.4 | NP_476843.2 | 12 - 13 | 3747 | 88 | ||
NM_057496.5 | NP_476844.2 | 14 - 15 | 4557 | 88 | ||
NM_057497.4 | NP_476845.2 | 13 - 14 | 5271 | 88 | ||
NM_166250.4 | NP_725723.2 | 11 - 12 | 2510 | 88 | ||
NM_166252.3 | NP_725725.1 | 11 - 12 | 2338 | 88 | ||
GenBank mRNA | BT029431.1 | - | 12 - 13 | 3441 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Drosophila melanogaster |