Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137458.3 | NP_611302.1 | 2 - 3 | 431 | 80 | |
NM_166274.2 | NP_725760.1 | 2 - 3 | 523 | 80 | ||
GenBank mRNA | BT133046.1 | - | 2 - 3 | 499 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |