Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166317.2 | NP_725841.1 | 1 - 2 | 339 | 70 | |
GenBank mRNA | AY058757.1 | - | 1 - 2 | 350 | 70 | |
U83596.1 | - | 1 - 2 | 310 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |