Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ena | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014536.2 | NP_001014536.1 | 4 - 5 | 809 | 68 | |
NM_001014537.3 | NP_001014537.1 | 4 - 5 | 733 | 68 | ||
NM_001144237.2 | NP_001137709.1 | 2 - 3 | 1470 | 68 | ||
NM_001299673.1 | NP_001286602.1 | 4 - 5 | 733 | 68 | ||
NM_166329.2 | NP_725857.1 | 4 - 5 | 733 | 68 | ||
NM_166330.2 | NP_725858.1 | 3 - 4 | 1169 | 68 | ||
NM_166331.3 | NP_725859.2 | 2 - 3 | 1017 | 68 | ||
GenBank mRNA | BT004488.1 | - | 4 - 5 | 720 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |