Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299678.1 | NP_001286607.1 | 14 - 15 | 5107 | 113 | |
NM_001299679.1 | NP_001286608.1 | 15 - 16 | 5395 | 113 | ||
NM_079067.4 | NP_523791.2 | 15 - 16 | 5401 | 113 | ||
NM_166336.2 | NP_725864.1 | 14 - 15 | 2416 | 113 | ||
NM_166337.2 | NP_725865.1 | 15 - 16 | 2974 | 113 | ||
NM_166338.3 | NP_725866.1 | 14 - 15 | 2404 | 113 | ||
GenBank mRNA | BT024976.1 | - | 14 - 15 | 2387 | 113 | |
BT133208.1 | - | 14 - 15 | 2402 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Drosophila melanogaster |