Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001299678.1 | NP_001286607.1 | 5 - 6 | 963 | 82 | |
NM_001299679.1 | NP_001286608.1 | 5 - 6 | 963 | 82 | ||
NM_079067.4 | NP_523791.2 | 5 - 6 | 963 | 82 | ||
NM_166336.2 | NP_725864.1 | 5 - 6 | 963 | 82 | ||
NM_166337.2 | NP_725865.1 | 5 - 6 | 963 | 82 | ||
NM_166338.3 | NP_725866.1 | 5 - 6 | 963 | 82 | ||
GenBank mRNA | BT024976.1 | - | 5 - 6 | 934 | 82 | |
BT133208.1 | - | 5 - 6 | 961 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |