Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cora | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166336.2 | NP_725864.1 | 9 - 10 | 1712 | 69 | |
NM_166337.2 | NP_725865.1 | 9 - 10 | 1700 | 69 | ||
GenBank mRNA | BT024976.1 | - | 9 - 10 | 1683 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |