Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hil | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137649.3 | NP_611493.2 | 4 - 5 | 862 | 76 | |
NM_166402.2 | NP_725980.1 | 4 - 5 | 925 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Drosophila melanogaster |