Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fem-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_137664.3 | NP_611508.1 | 5 - 6 | 2054 | 62 | |
NM_166408.2 | NP_725993.1 | 6 - 7 | 1861 | 62 | ||
GenBank mRNA | BT133040.1 | - | 5 - 6 | 1991 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |