Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Xpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_166429.3 | NP_726036.2 | 3 - 4 | 652 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Drosophila melanogaster |