Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sara | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079990.3 | NP_524729.2 | 2 - 3 | 1008 | 68 | |
GenBank mRNA | AF239997.1 | - | 2 - 3 | 985 | 68 | |
AJ310804.1 | - | 2 - 3 | 1013 | 68 | ||
BT058000.1 | - | 2 - 3 | 1016 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Drosophila melanogaster |