Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HmgD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001169773.2 | NP_001163244.1 | 2 - 3 | 290 | 70 | |
NM_001299767.1 | NP_001286696.1 | 2 - 3 | 818 | 70 | ||
NM_166479.2 | NP_726109.1 | 2 - 3 | 243 | 70 | ||
NM_166480.2 | NP_726110.1 | 2 - 3 | 242 | 70 | ||
GenBank mRNA | AY118333.1 | - | 1 - 2 | 215 | 70 | |
KX531848.1 | - | 1 - 2 | 149 | 70 | ||
M77023.1 | - | 2 - 3 | 247 | 70 | ||
X71138.1 | - | 242 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Drosophila melanogaster |