Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jbug | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001274210.1 | NP_001261139.1 | 3 - 4 | 1049 | 72 | |
NM_001274211.1 | NP_001261140.1 | 3 - 4 | 1049 | 72 | ||
NM_166546.3 | NP_726234.3 | 2 - 3 | 1088 | 72 | ||
NM_166547.2 | NP_726235.2 | 2 - 3 | 1088 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Drosophila melanogaster |