Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: inaD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001259541.1 | NP_001246470.1 | 7 - 8 | 1767 | 58 | |
NM_166566.1 | NP_726260.1 | 7 - 8 | 1731 | 58 | ||
GenBank mRNA | BT004473.1 | - | 7 - 8 | 1732 | 58 | |
BT120028.1 | - | 7 - 8 | 1694 | 58 | ||
U15803.1 | - | 7 - 8 | 1609 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Drosophila melanogaster |